(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)methanol 97%

Supplier: Ambeed
Warning

A119807-250MG A119807-1G
103799-842EA 125 USD
103799-842 77246-052 77311-948
(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)methanol 97%
(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.17 g/mol
Melting Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03659696
CAS Number: 274910-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR