1,4-Benzodioxane-5-boronic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2,3-Dihydrobenzo[b][1,4]dioxin-5-yl)boronic acid
A185646-250MG
A185646-100MG
103815-870EA
138
USD
103815-870
103846-440
77248-414
77746-201
1,4-Benzodioxane-5-boronic acid 95%
1,4-Benzodioxane-5-boronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BO₄ MW: 179.966 g/mol Boiling Pt: 372.6±52.0 °C (760 mm Hg) Flash Pt: 179.1 °C Storage Temperature: Freezer |
MDL Number:
MFCD03095151 CAS Number: 499769-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...