2,3-Difluoro-4-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Difluoro-4-methoxybenzeneboronic acid, (2,3-Difluoro-4-methoxyphenyl)boronic acid

A213314-10G A213314-250MG A213314-1G
103818-290EA 9 USD
103818-290 103848-996 77030-564 77030-566 77303-282
2,3-Difluoro-4-methoxyphenylboronic acid 97%
2,3-Difluoro-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
Boiling Pt: 304.3 °C (760 mm Hg)
Flash Pt: 137.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07368234
CAS Number: 170981-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR