(2,2-Dimethyl-1,3-dioxan-5-yl)methanol 98%

Supplier: Ambeed
Warning

A333396-1G A333396-5G A333396-100MG A333396-10G A333396-250MG A333396-100G
103825-640EA 9 USD
103825-640 103836-606 76839-612 76839-614 77032-932 77032-934 77032-936 77550-946
(2,2-Dimethyl-1,3-dioxan-5-yl)methanol 98%
(2,2-Dimethyl-1,3-dioxan-5-yl)methanol

Precaución: for Research Use Only

Formula: C₇H₁₄O₃
Boiling Pt: 207.7 °C (760 mm Hg)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04119722
CAS Number: 4728-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR