(2-(Trifluoromethyl)pyrimidin-5-yl)boronic acid 97%

Supplier: Ambeed
Warning

A191333-100MG A191333-250MG A191333-1G A191333-5G
103816-366EA 97 USD
103816-366 103846-968 77012-336 77012-338 77084-802 77084-804
(2-(Trifluoromethyl)pyrimidin-5-yl)boronic acid 97%
(2-(Trifluoromethyl)pyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₄BF₃N₂O₂
MW: 191.9 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10696932
CAS Number: 1308298-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR