(2-(Trifluoromethyl)pyridin-3-yl)methanamine 97%
Supplier: Ambeed
|
Danger
|
A394405-1G
A394405-5G
A394405-250MG
A394405-100MG
103826-942EA
167
USD
103826-942
103837-980
76841-390
76841-392
76876-824
77884-038
(2-(Trifluoromethyl)pyridin-3-yl)methanamine 97%
(2-(Trifluoromethyl)pyridin-3-yl)methanamine
Precaución: for Research Use Only
Formula:
C₇H₇F₃N₂ Boiling Pt: 194 °C (760 mm Hg) Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD10698574 CAS Number: 1056162-06-1 UN: 2924 ADR: 3,8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...