2-(Trifluoromethyl)benzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Hydroxymethyl)benzotrifluoride, (2-(Trifluoromethyl)phenyl)methanol

A566941-100G A566941-10G A566941-500G A566941-25G A566941-5G
103839-922EA 8 USD
103839-922 76696-260 76696-262 76696-264 76844-040 77307-830
2-(Trifluoromethyl)benzyl alcohol 97%
2-(Trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 191.3 °C (760 mm Hg)
Flash Pt: 92.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00004621
CAS Number: 346-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR