(2-(Methylthio)pyrimidin-4-yl)methanol 97%

Supplier: Ambeed
Warning

A123104-250MG A123104-1G A123104-5G
103800-340EA 61 USD
103800-340 77009-534 77312-460 77312-462
(2-(Methylthio)pyrimidin-4-yl)methanol 97%
(2-(Methylthio)pyrimidin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₈N₂OS
MW: 156.2 g/mol
Boiling Pt: 322.8 °C (760 mm Hg)
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD12964108
CAS Number: 102921-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR