2-(Methylthio)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-(Methylthio)phenyl)boronic acid, 2-(Methylthio)benzeneboronic acid

A151876-10G A151876-25G A151876-1G A151876-5G A151876-100G
103804-312EA 6 USD
103804-312 103842-462 76768-174 76768-176 77299-904 77318-046 77877-014
2-(Methylthio)phenylboronic acid 97%
2-(Methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₂S
Boiling Pt: 327.5 °C (760 mm Hg)
Melting Pt: 77…83 °C
Flash Pt: 151.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01318165
CAS Number: 168618-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR