(2-(Methoxymethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (2-methoxymethylphenyl)boronic acid, 2-(Methoxymethyl)phenylboronic acid, 2-(Methoxymethyl)benzeneboronic acid, 2-Methoxymethylphenylboronic acid

A196025-5G A196025-10G A196025-250MG A196025-1G
103816-802EA 10 USD
103816-802 103847-416 77085-372 77192-162 77192-164 77302-492
(2-(Methoxymethyl)phenyl)boronic acid 97%
(2-(Methoxymethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
Boiling Pt: 309.8 °C (760 mm Hg)
Melting Pt: 42…44 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412110
CAS Number: 126617-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR