2-(Hydroxymethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Hydroxymethyl)phenyl)boronic acid, 2-Hydroxymethylphenylboronic acid, 2-(Hydroxymethyl)benzeneboronic acid

A381258-1G A381258-25G A381258-5G A381258-100G A381258-10G
103826-694EA 6 USD
103826-694 103837-728 76841-044 76841-046 76841-048 77195-668 77195-670
2-(Hydroxymethyl)phenylboronic acid 98%
2-(Hydroxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Boiling Pt: 379.8 °C (760 mm Hg)
Melting Pt: 98…100 °C
Flash Pt: 183.5 °C
Storage Temperature: Ambient
MDL Number: MFCD02258966
CAS Number: 87199-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR