2-(Dimethylamino)-5-pyrimidineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-(Dimethylamino)pyrimidin-5-yl)boronic acid, 2-Dimethylaminopyrimidinyl-5-boronic acid

A337982-1G A337982-5G A337982-250MG
103825-760EA 20 USD
103825-760 103836-730 76839-748 76839-750 77672-728
2-(Dimethylamino)-5-pyrimidineboronic acid 98%
2-(Dimethylamino)-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₆H₁₀BN₃O₂
MW: 166.9735 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10696933
CAS Number: 756817-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR