(2-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%

Supplier: Ambeed
Warning

A126961-100MG A126961-250MG A126961-5G A126961-1G
103800-886EA 168 USD
103800-886 76983-486 76983-488 76983-490 77313-128
(2-(Difluoromethoxy)pyridin-3-yl)boronic acid 95%
(2-(Difluoromethoxy)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₆BF₂NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18261901
CAS Number: 1300750-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR