(2-(Aminomethyl)phenyl)methanol 97%

Supplier: Ambeed
Danger

A100100-1G A100100-5G
103795-252EA 29 USD
103795-252 103820-970 77296-566 77296-568
(2-(Aminomethyl)phenyl)methanol 97%
(2-(Aminomethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
Boiling Pt: 271 °C (760 mm Hg)
Flash Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09035091
CAS Number: 4152-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR