(2-((tert-Butoxycarbonyl)amino)pyrimidin-5-yl)boronic acid 95%
Supplier: Ambeed
|
Warning
|
A117708-250MG
A117708-1G
A117708-5G
103798-926EA
57
USD
103798-926
103824-814
77026-218
77311-590
77311-592
(2-((tert-Butoxycarbonyl)amino)pyrimidin-5-yl)boronic acid 95%
(2-((tert-Butoxycarbonyl)amino)pyrimidin-5-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₉H₁₄BN₃O₄ MW: 239.03 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16251533 CAS Number: 883231-25-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...