(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 98%

Supplier: Ambeed
Warning

Synonyms: tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

A184819-1G A184819-25G A184819-100G A184819-100MG A184819-10G A184819-250MG A184819-5G
103815-788EA 9 USD
103815-788 103846-354 77083-976 77083-978 77151-730 77151-732 77151-734 77151-736 77301-920
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 98%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
MW: 198.26 g/mol
Melting Pt: 74…76 °C
Flash Pt: 120.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01569250
CAS Number: 113451-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR