(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97%

Supplier: Ambeed
Warning

A330002-1G A330002-25G A330002-5G A330002-100MG A330002-10G A330002-250MG A330002-100G
103825-542EA 10 USD
103825-542 103836-500 76839-474 76839-476 76839-478 77114-860 77114-862 77114-864 77685-945
(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97%
(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one

Precaución: for Research Use Only

Formula: C₆H₇NO
Melting Pt: 94…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00211275
CAS Number: 130931-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR