(1S,4R)-4-Isopropenyl-1-methylcyclohex-2-en-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: (1S,4R)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-en-1-ol, (1S,4R)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-2-enol, cis-Isolimonenol

A111570-10G A111570-250MG A111570-25G A111570-1G A111570-5G A111570-500G A111570-100G
103797-862EA 7 USD
103797-862 103823-680 76821-866 76821-868 76821-870 77310-490 77310-492 77570-656 77683-021
(1S,4R)-4-Isopropenyl-1-methylcyclohex-2-en-1-ol 97%
(1S,4R)-4-Isopropenyl-1-methylcyclohex-2-en-1-ol

Precaución: For research use only.

Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 216.8 °C (760 mm Hg)
Melting Pt: 18 °C
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08460036
CAS Number: 22972-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR