1,3-Adamantanedicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Adamantanedicarboxylic Acid, 1,3-Dicarboxyadamantane, (1s,3s,5r,7r)-Adamantane-1,3-dicarboxylic acid

A109704-100G A109704-10G A109704-25G A109704-500G A109704-1G A109704-5G
103797-550EA 17 USD
103797-550 103823-356 76718-024 76718-026 76718-028 76718-030 77310-162 77310-164
1,3-Adamantanedicarboxylic acid 97%
1,3-Adamantanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₄
MW: 224.26 g/mol
Boiling Pt: 380.6 °C (760 mm Hg)
Melting Pt: 276…278 °C
Flash Pt: 198.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00167790
CAS Number: 39269-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR