Simvastatin 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3R,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate

A198975-100G A198975-10G A198975-250MG A198975-25G A198975-5G A198975-1G
103817-062EA 7 USD
103817-062 76721-374 76721-376 76721-378 76721-380 76721-382 77302-608
Simvastatin 98%
Simvastatin

Precaución: for Research Use Only

Formula: C25H38O5
MW: 418.57 g/mol
Boiling Pt: 564.9 °C (760 mm Hg)
Melting Pt: 139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072007
CAS Number: 79902-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR