(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A100682-100MG A100682-250MG A100682-5G
103795-574EA 42 USD
103795-574 103821-268 77148-176 77148-178 77296-806
(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 95%
(1S,3R)-Methyl 3-aminocyclopentanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂·ClH
MW: 179.645 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23106006
CAS Number: 180323-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR