Nomilin 98%

Supplier: Ambeed
Warning

Synonyms: (1S,3aS,4aR,4bR,6aR,11S,11aR,11bR,13aS)-1-(Furan-3-yl)-4b,7,7,11a,13a-pentamethyl-3,5,9-trioxohexadecahydrooxepino[4',3':3,4]benzo[1,2-f]oxireno[2,3-d]isochromen-11-yl acetate

A517627-1MG A517627-25MG A517627-10MG A517627-50MG
103792-754EA 26 USD
103792-754 103799-672 76726-188 76726-190 76843-578 76843-580
Nomilin 98%
Nomilin

Precaución: for Research Use Only

Formula: C₂₈H₃₄O₉
MW: 514.5600 g/mol
Boiling Pt: 657.7 °C (760 mm Hg)
Flash Pt: 351.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00075979
CAS Number: 1063-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR