(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 97%

Supplier: Ambeed
Warning

Synonyms: (1S,2S,3R,5S)-(+)-2,3-Pinanediol, (1S,2S,3R,5S)-(+)-Pinanediol

A148975-100G A148975-10G A148975-500G A148975-1G A148975-25G A148975-5G
103803-990EA 9 USD
103803-990 103842-110 76997-264 76997-266 76997-268 77299-698 77317-584 77317-586
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol 97%
(1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Melting Pt: 57…59 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00077851
CAS Number: 18680-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR