(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine 98%

Supplier: Ambeed
Warning

A269028-1G A269028-5G A269028-10G A269028-100MG A269028-250MG A269028-25G A269028-100G
103812-950EA 11 USD
103812-950 103852-854 77092-610 77092-612 77140-928 77571-666 77616-136 77616-138 77685-768
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine 98%
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MW: 142.242 g/mol
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09952017
CAS Number: 894493-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR