trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
(1S,2S)-(+)-trans-1,2-Bis(methylamino)cyclohexane, (1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
A262715-1G
A262715-10G
A262715-25G
A262715-5G
103812-576EA
7
USD
103812-576
103852-472
77092-094
77113-718
77304-858
77304-860
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine 97%
trans-(1S,2S)-N,N'-Dimethylcyclohexane-1,2-diamine
Precaución: for Research Use Only
Formula:
C₈H₁₈N₂ MW: 142.25 g/mol Boiling Pt: 186.8 °C (760 mm Hg) Melting Pt: 39…44 °C at N/A mmHg Flash Pt: 74 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00671528 CAS Number: 87583-89-9 UN: 3259 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...