(1S,2S)-Cyclohexane-1,2-diyldimethanol 97%

Supplier: Ambeed
Warning

A309945-250MG A309945-1G A309945-5G
103814-606EA 54 USD
103814-606 103835-786 76988-942 77306-010 77306-012
(1S,2S)-Cyclohexane-1,2-diyldimethanol 97%
(1S,2S)-Cyclohexane-1,2-diyldimethanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 270 °C (760 mm Hg)
Melting Pt: 60…63 °C
Flash Pt: 129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22572336
CAS Number: 3205-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR