(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1S)-trans-1,2-Diaminocyclohexane D-Tartrate, (1S,2S)-Cyclohexane-1,2-diamine (2S,3S)-2,3-dihydroxysuccinate
A592022-10G
A592022-250MG
103828-934EA
6
USD
103828-934
103840-080
77144-810
77686-502
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate 95%
(1S,2S)-(-)-1,2-Cyclohexanediamine-D-tartrate
Precaución: for Research Use Only
Formula:
C₆H₁₄N₂ MW: 264.28 g/mol Melting Pt: 280…284 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191980 CAS Number: 67333-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...