(1S,2S)-(+)-1,2-Diaminocyclohexane 96%
Supplier: Ambeed
|
Danger
|
A360682-1G
A360682-25G
A360682-5G
A360682-1000G
A360682-100G
A360682-10G
A360682-250MG
103826-266EA
6
USD
103826-266
103837-276
76840-488
76840-490
76840-492
77290-760
77290-762
77290-764
77290-766
(1S,2S)-(+)-1,2-Diaminocyclohexane 96%
(1S,2S)-(+)-1,2-Diaminocyclohexane
Precaución: for Research Use Only
Formula:
C₆H₁₄N₂ Boiling Pt: 104…110 °C (40 mm Hg) Melting Pt: 39…45 °C at N/A mmHg Flash Pt: 76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00062986 CAS Number: 21436-03-3 UN: 2735 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...