(1S,2S)-(+)-2-Aminocyclohexan-1-ol 98%

Supplier: Ambeed
Danger

Synonyms: (1S,2S)-(+)-2-Hydroxycyclohexan-1-amine, (1S,2S)-2-Aminocyclohexanol, (1S,2S)-(+)-1-Amino-2-hydroxycyclohexane

A114643-100G A114643-10G A114643-25G A114643-1G A114643-5G
103798-436EA 6 USD
103798-436 103824-280 76781-304 76781-306 76781-308 77297-314 77297-316
(1S,2S)-(+)-2-Aminocyclohexan-1-ol 98%
(1S,2S)-(+)-2-Aminocyclohexan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₃NO
Boiling Pt: 201.1 °C (760 mm Hg)
Melting Pt: 92…94 °C
Flash Pt: 75.4 °C
Storage Temperature: Ambient
MDL Number: MFCD08061326
CAS Number: 74111-21-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR