(1S,2S)-Boc-Acpc 95%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

A198799-5G A198799-100MG A198799-250MG
103817-046EA 79 USD
103817-046 103847-674 77085-696 77139-278 77139-280
(1S,2S)-Boc-Acpc 95%
(1S,2S)-Boc-Acpc

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD04974224
CAS Number: 143679-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR