(1S,2S)-(-)-1,2-Diphenylethylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-(-)-1,2-Diphenylethylenediamine, (1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, (S,S)-DPEN

A375912-1G A375912-25G A375912-5G A375912-100G A375912-10G
103826-574EA 5 USD
103826-574 103837-598 76840-896 76840-898 76840-900 77101-622 77101-624
(1S,2S)-(-)-1,2-Diphenylethylenediamine 98%
(1S,2S)-(-)-1,2-Diphenylethylenediamine

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂
MW: 212.29 g/mol
Boiling Pt: 353.9 °C (760 mm Hg)
Melting Pt: 83…85 °C
Flash Pt: 199.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00082751
CAS Number: 29841-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR