(1S,2R)-Boc-2-amino-1-cyclopentanecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: (1S,2R)-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid

A103190-1G A103190-5G A103190-250MG A103190-100MG
103796-502EA 78 USD
103796-502 103822-206 77309-082 77309-084 77592-094 77880-837
(1S,2R)-Boc-2-amino-1-cyclopentanecarboxylic acid 97%
(1S,2R)-Boc-2-amino-1-cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
Boiling Pt: 382.5 °C (760 mm Hg)
Flash Pt: 185.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09750528
CAS Number: 137170-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR