(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide 97%

Supplier: Ambeed
Danger

A268945-10G A268945-1G A268945-25G A268945-5G
103812-946EA 5 USD
103812-946 103852-850 77031-878 77031-880 77031-882 77092-602 77092-604 77092-606
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide 97%
(4aS,7R,8aS)-9,9-Dimethyltetrahydro-4H-4a-7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole-3,3-dioxide

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₃S
Boiling Pt: 317.6 °C (760 mm Hg)
Melting Pt: 172…174 °C
Flash Pt: 145.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00075319
CAS Number: 104322-63-6
UN: 1479
ADR: 5.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR