(1S)-(-)-Camphanic acid 96%

Supplier: Ambeed
Warning

Synonyms: (1S,4R)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid

A245550-100G A245550-10G A245550-1G A245550-25G A245550-5G
103811-654EA 10 USD
103811-654 103851-486 76692-360 76692-362 77090-740 77090-742 77090-744
(1S)-(-)-Camphanic acid 96%
(1S)-(-)-Camphanic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
Boiling Pt: 355.5 °C (760 mm Hg)
Melting Pt: 198…202 °C
Flash Pt: 142.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00044948
CAS Number: 13429-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR