(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%

Supplier: Ambeed
Danger

A222574-1G A222574-25G A222574-5G A222574-100G A222574-250MG A222574-500G A222574-10G
103818-986EA 6 USD
103818-986 103849-864 77088-472 77088-474 77088-476 77112-780 77112-782 77550-448 77685-598
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one 98%
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one

Precaución: for Research Use Only

Formula: C₆H₇NO
Melting Pt: 94…97 °C
Flash Pt: 167.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00211274
CAS Number: 79200-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR