(1R,3S,4S)-2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]heptane-3-carboxylic acid 97%

Supplier: Ambeed
Danger

A243729-100G A243729-10G A243729-1G A243729-5G
103811-540EA 28 USD
103811-540 103851-366 76987-678 76987-680 77304-278 77304-280
(1R,3S,4S)-2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]heptane-3-carboxylic acid 97%
(1R,3S,4S)-2-(tert-Butoxycarbonyl)-2-azabicyclo[2.2.1]heptane-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
MW: 241.28 g/mol
Melting Pt: 147…152 °C
Storage Temperature: Ambient
MDL Number: MFCD11099876
CAS Number: 291775-59-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR