(1R,3S)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 95%

Supplier: Ambeed
Warning

A205167-100MG A205167-250MG A205167-1G A205167-5G
103817-608EA 78 USD
103817-608 103848-274 77152-294 77152-296 77302-944 77302-946
(1R,3S)-Methyl 3-aminocyclopentanecarboxylate hydrochloride 95%
(1R,3S)-Methyl 3-aminocyclopentanecarboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD20925820
CAS Number: 180196-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR