(1R,3S)-3-Aminocyclopentanol hydrochloride 97%

Supplier: Ambeed
Warning

A647815-250MG A647815-25G
103840-384EA 58 USD
103840-384 76879-312 76879-314
(1R,3S)-3-Aminocyclopentanol hydrochloride 97%
(1R,3S)-3-Aminocyclopentanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11878010
CAS Number: 1279032-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR