(1R,3S)-3-Aminocyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

A159514-100MG A159514-250MG A159514-1G A159514-5G
103805-262EA 288 USD
103805-262 103843-482 76812-648 76812-650 77319-374 77319-376
(1R,3S)-3-Aminocyclopentanecarboxylic acid 95%
(1R,3S)-3-Aminocyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂
Boiling Pt: 264.7 °C (760 mm Hg)
Flash Pt: 113.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00211289
CAS Number: 71830-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR