(1R,2S)-cis-2-Aminocyclohexanol hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-2-Aminocyclohexanol hydrochloride

A261554-250MG A261554-1G A261554-100MG
103812-526EA 17 USD
103812-526 103852-416 76787-012 77092-020 77668-130
(1R,2S)-cis-2-Aminocyclohexanol hydrochloride 95%
(1R,2S)-cis-2-Aminocyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD11618002
CAS Number: 190792-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR