(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine (R)-2-hydroxy-2-phenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Ticagrelor Related Compound A

A311238-1G A311238-25G A311238-5G A311238-10G A311238-250MG A311238-100G
103814-658EA 6 USD
103814-658 103835-846 76838-638 76838-640 76838-642 76988-962 76988-964 77683-919
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine (R)-2-hydroxy-2-phenylacetate 98%
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropanamine (R)-2-hydroxy-2-phenylacetate

Precaución: For research use only.

Formula: C₁₇H₁₇F₂NO₃
MW: 321.31 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21362980
CAS Number: 376608-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR