(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1R,2S)-(+)-cis-1-Amino-2-indanol, (1R,2S)-(+)-cis-1-Aminoindan-2-ol
A186673-100G
A186673-10G
A186673-250MG
A186673-500G
A186673-1G
A186673-25G
A186673-5G
103815-956EA
9
USD
103815-956
103846-542
76998-864
76998-866
76998-868
76998-870
77084-218
77084-220
77084-222
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol 98%
(1R,2S)-1-Amino-2,3-dihydro-1H-inden-2-ol
Precaución: for Research Use Only
Formula:
C₉H₁₁NO MW: 149.19 g/mol Melting Pt: 118…121 °C Flash Pt: 129.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216656 CAS Number: 136030-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...