Isobornyl acrylate 95% stabilized

Supplier: Ambeed
Warning

Synonyms: (1R,2R,4R)-rel-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl acrylate, Acrylic acid Isobornyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A329828-100G A329828-500G A329828-25G A329828-1000G
103810-146EA 63 USD
103810-146 76803-030 76803-032 76839-472 77306-404
Isobornyl acrylate 95% stabilized
Isobornyl acrylate

Precaución: for Research Use Only

Formula: C₁₃H₂₀O₂
MW: 208.30 g/mol
Boiling Pt: 119…121 °C (15 mm Hg)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00080424
CAS Number: 5888-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR