Isobornyl methacrylate 95% stabilised

Supplier: Ambeed
Warning

Synonyms: rel-(1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate, (1R,2R,4R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-yl methacrylate(relative), Methacrylic acid Isobornyl Ester

Stabilization: 150-200 ppm Stabilised with 4-methoxyphenol (MEHQ)

A163225-100G A163225-10G A163225-500G A163225-25G
103808-926EA 59 USD
103808-926 76799-090 76799-092 76799-094 77320-050
Isobornyl methacrylate 95% stabilised
Isobornyl methacrylate

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Boiling Pt: 127…129 °C (15 mm Hg)
Melting Pt: 60 °C
Flash Pt: 107.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00081070
CAS Number: 7534-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR