tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 98%

Supplier: Ambeed
Danger

Synonyms: [(1R,2R)-2-Aminocyclohexyl]carbamic acid tert-Butyl ester, (1R,2R)-2-Amino-1-(Boc-amino)cyclohexane, (1R,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine, (1R,2R)-N-Boc-1,2-cyclohexanediamine, tert-Butyl [(1R,2R)-2-Aminocyclohexyl]carbamate, (1R,2R)-N1-Boc-1,2-cyclohexanediamine, (1R,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane

A172193-5G A172193-100MG A172193-10MG A172193-250MG A172193-50MG A172193-10G
103806-706EA 16 USD
103806-706 103844-990 77082-176 77138-442 77138-444 77138-446 77138-448 77615-442
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate 98%
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 322.1 °C (760 mm Hg)
Melting Pt: 114…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08726023
CAS Number: 146504-07-6
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR