(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine 95%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-N1,N1,N2,N2-Tetramethylcyclohexane-1,2-diamine

A380828-1G A380828-100MG A380828-250MG
103826-682EA 21 USD
103826-682 103837-716 76841-036 77101-692 77101-694
(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine 95%
(1R,2R)-1N,1N,2N,2N-Tetramethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₀H₂₂N₂
MW: 170.295 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03411208
CAS Number: 53152-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR