(1R,2R)-1,2-Cyclohexanedimethanol 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-1,2-Bis(hydroxymethyl)cyclohexane, (1R,2R)-Cyclohexane-1,2-diyldimethanol

A277107-10G A277107-1G
103813-332EA 6 USD
103813-332 103853-268 77032-074 77305-254
(1R,2R)-1,2-Cyclohexanedimethanol 97%
(1R,2R)-1,2-Cyclohexanedimethanol

Precaución: for Research Use Only

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 270.1 °C (760 mm Hg)
Melting Pt: 60…64 °C
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD16660860
CAS Number: 65376-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR