(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 95%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-Cyclohexane-1,2-diamine (2R,3R)-2,3-dihydroxysuccinate, (1R)-trans-1,2-Diaminocyclohexane L-Tartrate

A288729-100G A288729-10G A288729-25G A288729-1G
103813-806EA 7 USD
103813-806 103834-948 76802-344 76802-346 76837-514 77305-538
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate 95%
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 264.28 g/mol
Melting Pt: 273 °C
Storage Temperature: Ambient
MDL Number: MFCD00191979
CAS Number: 39961-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR