(1R,2R)-(-)-1,2-Diaminocyclohexane 96%

Supplier: Ambeed
Danger

A103470-100G A103470-10G A103470-250MG A103470-25G A103470-1G A103470-5G
103796-558EA 7 USD
103796-558 103822-264 77283-104 77283-106 77283-108 77283-110 77309-134 77309-136
(1R,2R)-(-)-1,2-Diaminocyclohexane 96%
(1R,2R)-(-)-1,2-Diaminocyclohexane

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
Boiling Pt: 163.773 °C (760 mm Hg)
Melting Pt: 43 °C at N/A mmHg
Flash Pt: 59.204 °C
Storage Temperature: Ambient
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR