(1R,2R)-2-Aminocyclopentanol hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (1R,2R)-2-aminocyclopentanol hydrochloride, (1|R|,2|R|)-2-Amino-1-hydroxycyclopentane Hydrochloride, trans-(1R,2R)-2-Aminocyclopentanol Hydrochloride, (1R,2R)-2-Amino-1-hydroxycyclopentane hydrochloride

A187080-100G A187080-10G A187080-250MG A187080-1G A187080-25G A187080-5G
103815-990EA 11 USD
103815-990 103846-582 76813-774 76813-776 76813-778 77084-272 77084-274 77084-276
(1R,2R)-2-Aminocyclopentanol hydrochloride 97%
(1R,2R)-2-Aminocyclopentanol hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁NO·HCl
MW: 137.61 g/mol
Flash Pt: 87.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09834692
CAS Number: 68327-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR